Learn More
Invitrogen™ GAB2 Polyclonal Antibody

Description
Immunogen sequence: ENSRSVAATI PRRNTLPAMD NSRLHRASSC ETYEYPQRGG ESAGRSAESM SDGVGSFLPG KMIVGRSDST NSEDNYVPMN PGSSTLLAME RAGDNSQSVY IPMSPGAHHF DSLGYPSTTL PVHRGPSRGS EIQPPPVNRN LKPDRKAKPT PLDLRNNTVI DELPFKSPIT KSWSRANHTF NSSSSQYCRP ISTQSITSTD SGDSEENYVP MQNPVSASPV PSGTNSPAPK KSTGSVDYLA LDFQPSSPSP HRKPSTSSVT SDEKVDYVQV DKEKTQALQN TMQEWTDVRQ SSEPSKGAKL; Positive Samples: HeLa, Mouse kidney; Cellular Location: Cell membrane, Cytoplasm.
Specifications
Specifications
| Antigen | GAB2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | GAB2 |
| Gene Accession No. | Q9UQC2, Q9Z1S8 |
| Gene Alias | AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100 |
| Gene Symbols | GAB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.