Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAB3 Polyclonal Antibody

Catalog No. PIPA5121972
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121972 20 μL
PIPA5119162 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121972 Supplier Invitrogen™ Supplier No. PA5121972
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GAB3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAB3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GAB3
Gene Accession No. Q8BSM5, Q8WWW8
Gene Alias 5930433H21Rik; DOS/Gab family member 3; GAB3; Gab3 scaffolding protein; GRB2 associated binding protein 3; GRB2-associated binder 3; GRB2-associated binding protein 3; GRB2-associated-binding protein 3; Growth factor receptor bound protein 2-associated protein 3
Gene Symbols GAB3
Host Species Rabbit
Immunogen Recombinant protein GRB2 Associated Binding Protein 3. The antigen corresponds to amino acid range 142-375 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139716, 210710
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.