Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R alpha 1 C-Terminus Antibody, R&D Systems™

Catalog No. PPS022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS022 Supplier R&D Systems Supplier No. PPS022
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GABA-AR alpha 1 Polyclonal specifically detects GABA-AR alpha 1 in Human, Mouse, Rat, Bovine, Canine, Primate, Zebrafish samples. It is validated for Western Blot.

Specifications

Antigen GABA-AR alpha 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias ECA4, EJM, EJM5GABA(A) receptor subunit alpha-1, GABAAR alpha 1, GABAARa1, GABRA1, gamma-aminobutyric acid (GABA) A receptor, alpha 1, gamma-aminobutyric acid receptor subunit alpha-1
Gene Symbols GABRA1
Host Species Rabbit
Immunogen Fusion protein from the cytoplasmic loop GABA A R alpha 1 subunit
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2554
Test Specificity Human, mouse, rat and bovine ∼51 kDa GABA A R alpha 1 subunit, C-Terminus
Target Species Human, Mouse, Rat, Bovine, Canine, Primate, Zebrafish
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.