Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-AR alpha 2 Antibody (S399-19), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259325IR
Description
GABA-AR alpha 2 Monoclonal specifically detects GABA-AR alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GABA-AR alpha 2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GABRA2 | |
| Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2 | |
| 100 μL | |
| Primary | |
| Detects 55kDa. Does not cross-react with GABA-A Receptor Alpha1. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S399-19 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2 | |
| Mouse | |
| Protein G purified | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2555 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction