Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GABA-A R delta |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GABA-A R delta Polyclonal specifically detects GABA-A R delta in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GABA-A R delta | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2563 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EIG10, EJM7, GABA(A) receptor subunit delta, GABA-A receptor, delta polypeptide, gamma-aminobutyric acid (GABA) A receptor, delta, gamma-aminobutyric acid receptor subunit delta, GEFSP5, MGC45284 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 330-430 of human GABA-A R delta (NP_000806.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title