Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R gamma 2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP149868
Description
GABA-A R gamma 2 Polyclonal specifically detects GABA-A R gamma 2 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
GABA-A R gamma 2 | |
Polyclonal | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_944494.1 | |
GABRG2 | |
Peptide with sequence C-NNATHLQERDE corresponding to internal region according to NP_000807.2, NP_944493.2. | |
0.1 mg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
2566 | |
Mouse, Rat | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CAE2, ECA2, GABA(A) receptor subunit gamma-2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize all reported isoforms: NP_944494.1; NP_000807.2; NP_944493.2. Note the aa numbers used in the name are from NP_944494.1. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction