Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R gamma 2 Antibody, R&D Systems™

Catalog No. PPS072
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS072 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS072 Supplier R&D Systems Supplier No. PPS072
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GABA-A R gamma 2 Polyclonal specifically detects GABA-A R gamma 2 in Human, Mouse, Rat, Bovine, Chicken, Primate, Zebrafish samples. It is validated for Western Blot.

Specifications

Antigen GABA-A R gamma 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias CAE2, ECA2, GABA(A) receptor subunit gamma-2, GABAAR gamma 2, GABAARg2, GABRG2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3
Gene Symbols GABRG2
Host Species Rabbit
Immunogen GABA-A R gamma 2
Purification Method Antigen Affinity-purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2566
Test Specificity Bovine, human, mouse, and rat ∼44-47 kDa gamma 2-subunit of the GABAA Receptor in Western blots.
Target Species Human, Mouse, Rat, Bovine, Chicken, Primate, Zebrafish
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.