Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA A Receptor alpha 6, Rabbit anti-Rat, Bovine, Polyclonal, Millipore Sigma™

Catalog No. AB5610I25UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5610I25UG 25 μg
AB5610I100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB5610I25UG Supplier MilliporeSigma Supplier No. AB5610I25UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GABA A Receptor alpha 6, Cat. No. AB5610-I, is a highly specific rabbit polyclonal antibody that targets Gamma-aminobutyric acid type A receptor alpha6 subunit and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

Gamma-aminobutyric acid type A receptor alpha 6 subunit (UniProt: E1BE96; also known as GABA A receptor alpha 6) is encoded by the GABRA6 gene (Gene ID: 539194) in bovine. GABA is the major inhibitory neurotransmitter in the central nervous system that inhibits the generation of the action potential of the neuron. It is involved in the pathogenesis of certain neurological and psychiatric disorders. GABA is produced from glutamic acid in a reaction catalyzed by glutamic acid decarboxylase. GABA interacts with the GABAA and GABAB receptors, which are widely distributed throughout the nervous system and in a variety of cell types. They differ in their pharmacological, electrophysiological, and biochemical properties. GABAA-receptor are pentameric membrane proteins that mediate an increase in membrane conductance with an equilibrium potential near the resting level of 70 mV. This conductance increase often is accompanied by a membrane hyperpolarization, which later results in a reduction in the probability of action potential initiation. The reduction in membrane resistance is accomplished by the GABA-dependent facilitation of chloride ion influx. Mos GABA A receptors contain one type of alpha and beta subunit, and a single gamma polypeptide in a ratio of 2:2:1. GABA A receptor alpha 6 subunit is synthesized with a signal peptide (aa 1-19), which is cleaved off in the mature form. It contains four transmembrane domains (aa 240-262, 274-293, 305-328, and 423-441).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA A Receptor alpha 6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected GABA A Receptor alpha 6 in mouse cerebellum tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Symbols GABRA6
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of bovine GABA A receptor alpha 6 subunit.
Molecular Weight of Antigen ∽51 kDa observed; 51.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NM_000811.2
Test Specificity This rabbit polyclonal antibody detects GABA A receptor alpha 6 subunit. It targets an epitope with in 16 amino acids from the N-terminal region.
Target Species Rat, Bovine
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.