Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA A Receptor beta 2 + 3 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981917
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1917 Supplier Bioss Supplier No. BS12066R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gamma-aminobutyric acid type A (GABAA) receptors are members of the neurotransmitter ligand-gated ion channels that mediate neuronal inhibition on binding GABA. The effects of GABA on GABAA receptors are modulated by a range of therapeutically important drugs, including barbiturates, anaesthetics and benzodiazepines.

Specifications

Antigen GABA A Receptor beta 2 + 3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GABRB2
Gene Accession No. P47870, P63137, P63138
Gene Symbols GABRB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GABA A Receptor beta 2 + 3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14401, 25451, 2561
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.