Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-AR alpha 5 Antibody (S415-24), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259700B
Description
GABA-AR alpha 5 Monoclonal specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GABA-AR alpha 5 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GABRA5 | |
Fusion protein amino acids 368-419 (last cytoplasmic domain before extracellular C-terminus) of human GABA A Receptor Alpha5. | |
100 μL | |
Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
2558 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S415-24 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects approx 30kDa. No cross-reactivity against KChIP2, KChIP3, or KChIP4. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction