Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-B R1 Antibody (S93A-49), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259336R
Description
GABA-B R1 Monoclonal specifically detects GABA-B R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GABA-B R1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GABBR1 | |
Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1 | |
100 μL | |
Primary | |
Detects 115kDa. No cross-reactivity against GABA(B)R2. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S93A-49 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor | |
Mouse | |
Protein G purified | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
2550 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction