Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-B R1 Antibody (SR1045), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32180250UL
Description
GABA-B R1 Monoclonal antibody specifically detects GABA-B R1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
GABA-B R1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1045 | |
Western Blot 1:500-1:2000 | |
FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor | |
A synthesized peptide derived from human GABA-B R1 (Uniprot #: Q9UBS5) | |
50 μg | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
2550 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction