Learn More
GABA-B R2 Antibody (S81-2), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259335B
Catalog No. NBP259335B
Only null left
Add to Cart
Description
Description
GABA-B R2 Monoclonal specifically detects GABA-B R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GABA-B R2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
GABBR2
Fusion protein amino acids 861-912 of rat GABA(B)R2
100 μL
Primary
Detects 105kDa. No cross-reactivity against GABA(B)R1.
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S81-2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099
Mouse
Protein G purified
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
9568
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.