Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA B Receptor 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207111
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7111 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7111 Supplier Fabgennix Inc Supplier No. GABAB2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABA B Receptor 2 Polyclonal antibody specifically detects GABA B Receptor 2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen GABA B Receptor 2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene GABBR2
Gene Accession No. O75899, O88871, Q80T41
Gene Alias FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on human gamma-aminobutyric acid A receptor beta 2 protein. Will label various GABA B subunit 2 variants.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242425, 83633, 9568
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.