Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA Transporter-3, CT Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$191.34 - $481.79
Specifications
Antigen | GABA Transporter-3, CT |
---|---|
Concentration | 1.0 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AB1574I25UG
|
MilliporeSigma
AB1574I25UG |
25 μg |
Each for $191.34
|
|
|||||
AB1574I100
|
MilliporeSigma
AB1574I100UG |
100 μg |
Each for $481.79
|
|
|||||
Description
GABA Transporter-3, CT Polyclonal antibody specifically detects GABA Transporter-3, CT in Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).Specifications
GABA Transporter-3, CT | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat | |
KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal, cytoplasmic domain of rat GABA Transporter 3. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
1.0 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title