Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA Transporter 3/GAT 3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13260R

Description
Slc6a11 encode GABA transporters that are sodium- and chloride-dependent members of the solute carrier family 6 (SLC6) and mediate the rapid removal of GABA and maintain low extracellular levels. Four 12-TM domain transporters have been identified and localized to neurons and glia. The protein encoded by the Slc6a11 gene is a sodium-dependent transporter that uptakes gamma-aminobutyric acid (GABA), an inhibitory neurotransmitter, which ends the GABA neurotransmission. Defects in the Slc6a11 gene may result in epilepsy, behavioral problems, or intellectual problems. Two transcript variants encoding different isoforms have been found for the Slc6a11 gene. Diseases associated with SLC6A11 include Tricuspid Valve Stenosis and Pulmonary Valve Insufficiency.Specifications
GABA Transporter 3/GAT 3 | |
Polyclonal | |
Unconjugated | |
SLC6A11 | |
BOS_21188; D930045G19Rik; E130202I16Rik; GABA transporter 3; GABA transporter GAT-3; Gabt3; Gabt4; gamma-aminobutyric acid (GABA-A) transporter 4; Gat3; Gat-3; Gat4; Gat-4; Gat-b; LOC100348900; SLC6A11; Sodium- and chloride-dependent GABA transporter 3; sodium- and chloride-dependent GABA transporter 3-like; Sodium- and chloride-dependent GABA transporter 4; solute carrier family 6 (neurotransmitter transporter), member 11; solute carrier family 6 (neurotransmitter transporter, GABA), member 11; Solute carrier family 6 member 11 | |
Rabbit | |
Protein A | |
RUO | |
6538 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P48066 | |
SLC6A11 | |
KLH conjugated synthetic peptide derived from human GABA Transporter 3/GAT 3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction