Learn More
GABAA R gamma 2 C-Terminus Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | GABA-A R gamma 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 dilution |
| Gene Alias | CAE2, ECA2, GABA(A) receptor subunit gamma-2, GABAAR gamma 2, GABAARg2, GABRG2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3 |
| Gene Symbols | GABRG2 |
| Host Species | Rabbit |
| Immunogen | Fusion protein from the cytosolic loop of the rat GABA A R gamma 2 subunit, C-Terminus |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.