Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabapentin, Tocris Bioscience™

Catalog No. p-4783472
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Increases brain GABA; anticonvulsant

Anticonvulsant with several possible mechanisms of action. Increases GABA in the brain and binds to a novel site associated with voltage-sensitive Ca2+ channels. Prevents neuronal death and is antinociceptive and anxiolytic.

Chemical Identifiers

CAS 60142-96-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
InChI Key UGJMXCAKCUNAIE-UHFFFAOYSA-N
Synonym gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
PubChem CID 3446
ChEBI CHEBI:42797
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CN

Specifications

Quantity 10 mg
Formula Weight 171.24
Percent Purity >99%
Chemical Name or Material Gabapentin
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.