Learn More
Invitrogen™ GABARAP Polyclonal Antibody

Description
PA1-4146 detects GABA Receptor Associated Protein Subunit 2 in human and rat samples. PA1-4146 has been successfully used Western blot procedures. By Western blot this antibody detects a ~105 kDa protein representing GABA Receptor Associated Protein Subunit 2 in brain lysate. The PA1-4146 immunogen is a synthetic peptide corresponding to residues T(42) R G A P R P P P S S P P(54) of the 105 kDa rat GABA R2 protein.
Specifications
Specifications
| Antigen | GABARAP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GABARAP |
| Gene Accession No. | O95166, P60517 |
| Gene Alias | ATG8A; FLC3B; FLJ25768; GABA type A receptor-associated protein; GABA(A) receptor-associated protein; GABA-A receptor-associated protein; Gabarap; GABARAP a; GABARAP-a; gamma-aminobutyric acid receptor associated protein; gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; HT004; hypothetical protein; MGC120154; MGC120155; MM46 |
| Gene Symbols | GABARAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.