Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABARAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14147
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14147 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14147 Supplier Invitrogen™ Supplier No. PA14147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4147 detects GABA Receptor Associated Protein in human, rat, mouse, bovine, and non-human primate samples. PA1-4147 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~116 kDa protein representing GABA Receptor Associated Protein in brain lysate. The PA1-4147 immunogen is a synthetic peptide corresponding to residues P(33) H L P R P H P R V P P H P S S E R R A V Y(54) of the mouse and rat GABA R1 subunit.

Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system. It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. GABARAP (GABAA receptor-associated protein) links GABAA receptors to the cytoskeleton and may play a role in intracellular transport of GABAA receptors and its interaction with the cytoskeleton. GABARAP belongs to the MAP1 or ATG8 like family and recent studies show that MAPK15/ERK8 is acting through interaction with ATG8 family proteins to regulate autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABARAP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GABARAP
Gene Accession No. O95166, P60517, Q9DCD6, Q9GJW7
Gene Alias ATG8A; FLC3B; FLJ25768; GABA type A receptor-associated protein; GABA(A) receptor-associated protein; GABA-A receptor-associated protein; Gabarap; GABARAP a; GABARAP-a; gamma-aminobutyric acid receptor associated protein; gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; HT004; hypothetical protein; MGC120154; MGC120155; MM46
Gene Symbols GABARAP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P(33) H L P R P H P R V P P H P S S E R R A V Y(54) of the mouse and rat GABA R1 subunit.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11337, 327715, 56486, 58974
Target Species Human, Mouse, Rat, Monkey, Bovine
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.