Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAPL1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E10F6, Proteintech
SDP

Catalog No. 501736884 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6884 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6884 Supplier Proteintech Group Inc Supplier No. 664581IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.

Specifications

Antigen GABARAPL1
Applications Western Blot
Classification Monoclonal
Clone 1E10F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GABARAPL1
Gene Accession No. Q0VGK0, Q8R3R8, Q9H0R8
Gene Alias APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1, GEC 1, GEC1
Gene Symbols Gabarapl1
Host Species Mouse
Immunogen GABARAPL1 Fusion Protein Ag1473
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100462751, 23710, 57436, 689161
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.