Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABARAPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544334
Description
Peptide sequence: KAPKARVPDL DKRKYLVPSD LTVGQFYFLI RKRIHLRPED ALFFFVNNTI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
Specifications
GABARAPL1 | |
Polyclonal | |
Unconjugated | |
GABARAPL1 | |
3110025G09Rik; 9130422N19Rik; AI196471; Apg8l; APG8LIKE; APG8-LIKE; ATG8; ATG8B; Atg8l; Early estrogen-regulated protein; GABA type A receptor associated protein like 1; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABA(A)-RAP; GABARAPL1; GABARAPL1-a; gamma-aminobutyric acid (GABA(A)) receptor-associated protein-like 1; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1; gamma-aminobutyric acid receptor-associated protein-like 1; gamma-aminobutyric acid-receptor-associated protein; Gec1; GEC-1; GECI; Glandular epithelial cell protein 1; MDBK gamma-aminobutyric acid-receptor-associated protein; MNCb-0091 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23710 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0R8 | |
GABARAPL1 | |
Synthetic peptide directed towards the middle region of human GABARAPL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction