Learn More
GABARAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GABARAPL1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2207
![]() ![]() |
Proteintech Group Inc
187211AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to GABARAPL1, and does not recognize the recombit GABARAP and GABARAPL2.
Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.Specifications
GABARAPL1 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
23710, 57436, 689161 | |
Peptide | |
Primary | |
-20°C | |
GABARAPL1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1, GEC 1, GEC1 | |
Gabarapl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.