Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAPL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988220
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8220 Supplier Bioss Supplier No. BS8035R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.

Specifications

Antigen GABARAPL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GABARAPL1
Gene Accession No. Q9H0R8
Gene Alias 3110025G09Rik; 9130422N19Rik; AI196471; Apg8l; APG8LIKE; APG8-LIKE; ATG8; ATG8B; Atg8l; BOS_5888; Early estrogen-regulated protein; GABA type A receptor associated protein like 1; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABARAPL1; GABARAPL1-a; gamma-aminobutyric acid (GABA(A)) receptor-associated protein-like 1; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1; Gamma-aminobutyric acid receptor-associated protein-like 1; Gec1; GEC-1; GECI; Glandular epithelial cell protein 1; LOC100340751; MDBK gamma-aminobutyric acid-receptor-associated protein; MNCb-0091
Gene Symbols GABARAPL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GABARAPL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23710
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.