Learn More
Invitrogen™ GABARAPL1 Recombinant Rabbit Monoclonal Antibody (5P10L6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558105

Description
Immunogen sequence: MKFQYKEDHP FEYRKKEGEK IRKKYPDRVP VIVEKAPKAR VPDLDKRKYL VPSDLTVGQF YFLIRKRIHL RPEDALFFFV NNTIPPTSAT MGQLYEDNHE.
GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
Specifications
GABARAPL1 | |
Recombinant Monoclonal | |
1.74 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
GABARAPL1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GABARAPL1 (NP_113600.1). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
5P10L6 | |
Unconjugated | |
GABARAPL1 | |
3110025G09Rik; 9130422N19Rik; AI196471; Apg8l; APG8LIKE; APG8-LIKE; ATG8; ATG8B; Atg8l; Early estrogen-regulated protein; GABA type A receptor associated protein like 1; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABA(A)-RAP; GABARAPL1; GABARAPL1-a; gamma-aminobutyric acid (GABA(A)) receptor-associated protein-like 1; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1; gamma-aminobutyric acid receptor-associated protein-like 1; gamma-aminobutyric acid-receptor-associated protein; Gec1; GEC-1; GECI; Glandular epithelial cell protein 1; MDBK gamma-aminobutyric acid-receptor-associated protein; MNCb-0091 | |
Rabbit | |
Protein A | |
RUO | |
23710, 57436, 689161 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.