Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABARAPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587030
Description
Purity is > 95% by SDS-PAGE.
Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Specifications
GABARAPL2 | |
Polyclonal | |
Unconjugated | |
GABARAPL2 | |
0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABA type A receptor associated protein like 2; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gabarapl2; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE16; GATE-16; Gef2; GEF-2; Genera; general protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein; zgc:92319 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11345, 64670, 93739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P60520, P60521, P60522 | |
GABARAPL2 | |
Recombinant fusion protein of human GABARAPL2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction