Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABARAPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114488
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114488 Supplier Invitrogen™ Supplier No. PA5114488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: Human Brain Tissue Lysate.

Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABARAPL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GABARAPL2
Gene Accession No. P60520, P60521, P60522
Gene Alias 0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABA type A receptor associated protein like 2; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gabarapl2; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE16; GATE-16; Gef2; GEF-2; Genera; general protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein; zgc:92319
Gene Symbols GABARAPL2
Host Species Rabbit
Immunogen GABARAPL2 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human GABARAPL2. The immunogen is located within the last 50 amino acids of GABARAPL2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11345, 64670, 93739
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.