Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABARAPL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572222
Description
GABARAPL2 Polyclonal Antibody for Western Blot
Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Specifications
GABARAPL2 | |
Polyclonal | |
Unconjugated | |
GABARAPL2 | |
0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABA type A receptor associated protein like 2; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gabarapl2; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE16; GATE-16; Gef2; GEF-2; Genera; general protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein; zgc:92319 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
403036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.61 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
GABARAPL2 | |
KLH conjugated synthetic peptide between 15-48 amino acids from the N-terminal region of DANRE gabarapl2. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction