Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAPL2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC22725UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC227-25UL 25 μL
ABC227-100UL 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC227-25UL Supplier MilliporeSigma Supplier No. ABC22725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GABARAPL2 is a rabbit polyclonal antibody that detects Gamma-aminobutyric acid receptor-associated protein-like 2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABARAPL2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols GABARAPL2,FLC3A,GEF2
Host Species Rabbit
Immunogen GST-tagged full length human recombinant GABA(A) receptor-associated protein-like 2 (GABARAPL2).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects GABA(A) receptor-associated protein-like 2 (GABARAPL2) in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.