Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146248
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146248 Supplier Invitrogen™ Supplier No. PA146248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: MDA-MB-231, human brain protein. Immunogen sequence: PHLPRPHPRV PPHPSSERRA VY.

Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABBR1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GABBR1
Gene Accession No. Q9UBS5
Gene Alias bM573K1.1; bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABAB; GABA-B receptor 1; GABA-B receptor, R1 subunit; GABAB1; Gaba-b1; GABAbR1; GABA-BR1; GABA-B-R1; Gababr1 receptor; Gaba-b-r1a; Gaba-br1a receptor; GABA-BR1b receptor; Gabbr1; GABBR1-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 1; gamma-aminobutyric acid (GABA) B receptor, 1; gamma-aminobutyric acid (GABA-B) receptor, 1; gamma-aminobutyric acid type B receptor subunit 1; GB1; GBR1; GPRC3A; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor
Gene Symbols GABBR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 33-54 of the GABABR1 subunit.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.