Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABBR2 Monoclonal Antibody (N81/2), PerCP
GREENER_CHOICE

Catalog No. PIMA545535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545535 Supplier Invitrogen™ Supplier No. MA545535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45535 was sufficient for detection of GABA(B)R2 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 105kDa. No cross-reactivity against GABA(B)R1. This antibody was formerly sold as clone S81-2.

The multi-pass membrane protein, GABBR2, belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABBR2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N81/2
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene GABBR2
Gene Accession No. O75899, O88871, Q80T41
Gene Alias FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1
Gene Symbols GABBR2
Host Species Mouse
Immunogen Fusion protein amino acids 861-912 of rat GABA(B)R2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242425, 83633, 9568
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.