Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABBR2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA110032
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA110032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA110032 Supplier Thermo Scientific Supplier No. PA110032

Rabbit Polyclonal Antibody

This antibody recognizes the C-terminus of gamma-aminobutyric acid (GABA) B receptor 2 (epitope: the last 23 aa).

GABBR2 Polyclonal antibody specifically detects GABBR2 in Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABBR2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene GABBR2
Gene Accession No. O88871, Q80T41
Gene Alias FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1
Gene Symbols GABBR2
Host Species Rabbit
Immunogen Synthetic peptide (coupled with THG) derived from the last 23 aa of mouse GABA B receptor 2. 100% homology with human GB2.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242425, 83633
Target Species Mouse, Rat
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.