Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527795
Description
Recommended positive controls: Raji, K562. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (88%), Chicken (87%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype. Two transcript variants encoding the same protein have been found for this gene.
Specifications
GABPA | |
Polyclonal | |
Unconjugated | |
GABPA | |
E4TF1-60; E4TF1A; GA binding protein transcription factor alpha subunit; GA binding protein transcription factor alpha subunit 60kDa; GA binding protein transcription factor, alpha subunit; GA binding protein transcription factor, alpha subunit 60kDa; GA repeat binding protein alpha; GA repeat binding protein, alpha; GA-binding protein alpha chain; GA-binding protein alpha-subunit; GABP subunit alpha; GABPA; GABPalpha; human nuclear respiratory factor-2 subunit alpha; NFT2; NRF2; Nrf2a; Nrf-2alpha; nuclear respiratory factor 2 alpha; nuclear respiratory factor 2 alpha subunit; nuclear respiratory factor 2 subunit alpha; RCH04A07; Transcription factor E4TF1-60 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.02 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q06546 | |
GABPA | |
Recombinant protein encompassing a sequence within the N-terminus region of human GABPA. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction