Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583032 Supplier Invitrogen™ Supplier No. PA583032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDGDMICKVQ GKRFVYKFVC DLKTLIGYSA AELNRLVTEC EQKKLAKMQL HGIAQPVTAV ALATASLQTE KDN Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 89%.

This gene encodes one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABPA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GABPA
Gene Accession No. Q06546
Gene Alias E4TF1-60; E4TF1A; GA binding protein transcription factor alpha subunit; GA binding protein transcription factor alpha subunit 60kDa; GA binding protein transcription factor, alpha subunit; GA binding protein transcription factor, alpha subunit 60kDa; GA repeat binding protein alpha; GA repeat binding protein, alpha; GA-binding protein alpha chain; GA-binding protein alpha-subunit; GABP subunit alpha; GABPA; GABPalpha; human nuclear respiratory factor-2 subunit alpha; NFT2; NRF2; Nrf2a; Nrf-2alpha; nuclear respiratory factor 2 alpha; nuclear respiratory factor 2 alpha subunit; nuclear respiratory factor 2 subunit alpha; RCH04A07; Transcription factor E4TF1-60
Gene Symbols GABPA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GABPA. Recombinant protein control fragment (Product #RP-95878).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2551
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.