Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABPA Polyclonal Antibody, Huabio

Catalog No. 502544585
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4585 Supplier HUABIO RESEARCH INC Supplier No. HA500187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABPA Polyclonal antibody specifically detects GABPA in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen GABPA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene GABPA
Gene Accession No. Q00422, Q06546
Gene Alias E4TF1-60; E4TF1A; GA binding protein transcription factor alpha subunit; GA binding protein transcription factor alpha subunit 60kDa; GA binding protein transcription factor, alpha subunit; GA binding protein transcription factor, alpha subunit 60kDa; GA repeat binding protein alpha; GA repeat binding protein, alpha; GA-binding protein alpha chain; GA-binding protein alpha-subunit; GABP subunit alpha; GABPA; GABPalpha; human nuclear respiratory factor-2 subunit alpha; NFT2; NRF2; Nrf2a; Nrf-2alpha; nuclear respiratory factor 2 alpha; nuclear respiratory factor 2 alpha subunit; nuclear respiratory factor 2 subunit alpha; RCH04A07; Transcription factor E4TF1-60
Host Species Rabbit
Immunogen Recombinant protein within human GABPA aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14390, 2551
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.