Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABPB1 Monoclonal Antibody (OTI3F1), TrueMAB™, OriGene
SDP

Catalog No. 501683940
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3940 Supplier OriGene Supplier No. TA811260
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.

Specifications

Antigen GABPB1
Applications Western Blot
Classification Monoclonal
Clone OTI3F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GABPB1
Gene Accession No. Q06547
Gene Alias BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
Gene Symbols GABPB1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 128-360 of human GABPB1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2553
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.