Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABPB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592021 Supplier Invitrogen™ Supplier No. PA592021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNSHRATANT EEIIEGNSVD SSIQQVMGSG GQRVITIVTD GVPLGNIQTS IPTGGIGQPF IVTVQDGQQV LTVPAGKVAE ETVIKEEEEE KLPLTKKPRI GEKTNSVEES KEGNERELLQ QQLQEANRRA QEYRHQLLKK EQEAEQYRLK LEAIARQQPN GVDFTMVEEV AEVDAVVVTE GELEERETKV TGSAGTTEPH TRVSMATVSS; Positive Samples: Mouse thymus, Mouse spleen, Mouse kidney, Rat thymus; Cellular Location: Nucleus.

GABPB2 (GA binding protein transcription factor, beta subunit 2) is a protein-coding gene. GO annotations related to this gene include protein homodimerization activity and transcription regulatory region DNA binding. An important paralog of this gene is ANKRD2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABPB2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gabpb2
Gene Accession No. P81069
Gene Alias 1810015F01Rik; 5830427M07; 9430006E19Rik; A430024B14Rik; AV050852; BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor beta subunit 2; GA binding protein transcription factor subunit beta 2; GA binding protein transcription factor, alpha subunit pseudogene; GA binding protein transcription factor, beta subunit 1; GA binding protein transcription factor, beta subunit 2; GA repeat binding protein, beta 2; GA-binding protein beta-2-1; GA-binding protein subunit beta-1; GA-binding protein subunit beta-2; GABP subunit beta-1; GABP subunit beta-2; GABP subunit beta-2-1; GABP2; GABPAP; GABPB; GABPB1; GABPB-1; Gabpb2; GABPB-2; Gabpb2-1; LOW QUALITY PROTEIN: GA-binding protein subunit beta-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1565234; RP11-68I18.1; Transcription factor E4TF1-47; transcription factor E4TF1-53
Gene Symbols Gabpb2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 239-448 of human GABPB2 (NP_6532191).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213054, 295263
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.