Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABPB2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986360
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6360 Supplier Bioss Supplier No. BS5140R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABPB2 (GA binding protein transcription factor, beta subunit 2) is a protein-coding gene. GO annotations related to this gene include protein homodimerization activity and transcription regulatory region DNA binding. An important paralog of this gene is ANKRD2.

Specifications

Antigen GABPB2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gabpb2
Gene Accession No. Q8TAK5
Gene Alias 1810015F01Rik; 5830427M07; 9430006E19Rik; A430024B14Rik; AV050852; BABPB2; BOS_2948; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GA binding protein transcription factor beta subunit 2; GA binding protein transcription factor subunit beta 2; GA binding protein transcription factor, alpha subunit pseudogene; GA binding protein transcription factor, beta subunit 1; GA binding protein transcription factor, beta subunit 2; GA repeat binding protein, beta 2; GA-binding protein beta-2-1; GA-binding protein subunit beta-2; GABP subunit beta-2; GABP subunit beta-2-1; GABP2; GABPAP; GABPB; GABPB1; Gabpb2; GABPB-2; Gabpb2-1; NRF2B1; NRF2B2; RGD1565234; RP11-68I18.1
Gene Symbols Gabpb2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GABPB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126626
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.