Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA1 Monoclonal Antibody (N95/35), APC
GREENER_CHOICE

Catalog No. PIMA545485
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545485 Supplier Invitrogen™ Supplier No. MA545485
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45485 was sufficient for detection of alpha1 GABA receptor in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 55kDa. No cross-reactivity against GABA-A-R-Alpha 2 or -Alpha3.

Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N95/35
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene GABRA1
Gene Accession No. P14867, P62812, P62813
Gene Alias ECA4; EIEE19; EJM; EJM5; GABA alpha subunit; GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; GABAA alpha 1; GABAAR alpha1; GABRA 1; GABRA1; Gabra-1; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor alpha 1 variant 1; gamma-aminobutyric acid A receptor alpha 1 variant 2; gamma-aminobutyric acid A receptor alpha 1 variant 3; gamma-aminobutyric acid A receptor alpha 1 variant 4; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid receptor subunit alpha-1; gamma-aminobutyric acid receptor subunit alpha-1-like; gamma-aminobutyric acid type A receptor alpha1 subunit; gamma-aminobutyric acid type A receptor subunit alpha1; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-1
Gene Symbols GABRA1
Host Species Mouse
Immunogen Fusion protein amino acids 355-394 of human GABA-A-R-Alpha1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14394, 2554, 29705
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.