Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114315 Supplier Invitrogen™ Supplier No. PA5114315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABRA1 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene GABRA1
Gene Accession No. P14867, P62813
Gene Alias ECA4; EIEE19; EJM; EJM5; GABA alpha subunit; GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; GABAA alpha 1; GABAAR alpha1; GABRA 1; GABRA1; Gabra-1; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor alpha 1 variant 1; gamma-aminobutyric acid A receptor alpha 1 variant 2; gamma-aminobutyric acid A receptor alpha 1 variant 3; gamma-aminobutyric acid A receptor alpha 1 variant 4; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid receptor subunit alpha-1; gamma-aminobutyric acid receptor subunit alpha-1-like; gamma-aminobutyric acid type A receptor alpha1 subunit; gamma-aminobutyric acid type A receptor subunit alpha1; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-1
Gene Symbols GABRA1
Host Species Rabbit
Immunogen Synthetic peptide within rat GABRA1 aa 11-60.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2554, 29705
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.