Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14713 Supplier Invitrogen™ Supplier No. PA14713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4713 is specific for the ~51k alpha-2-subunit of the GABAA receptor. In Western blot, labeling is absent in alpha-2-subunit knockout animals. This antibody is predicted to react with bovine, canine, human, non-human primate and Zebrafish based on 100% sequence homology.

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene GABRA2
Gene Accession No. P23576, P26048
Gene Alias C630048P16Rik; FLJ97076; GABA(A) receptor; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; GABAARalpha2; Gabra2; Gabra-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; gamma-aminobutyric acid receptor subunit alpha-2; gamma-aminobutyric acid type A receptor alpha2 subunit
Gene Symbols GABRA2
Host Species Rabbit
Immunogen Fusion protein from the cytoplasmic loop of the alpha 2 subunit of rat GABRA2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14395, 289606
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.