Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABRA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109751
Description
Immunogen sequence: ARNSLPKVAY ATAMDWFIAV CYAFVFSALI EFATVNYFTK RGWAWDGKSV VNDKKKEKAS VMIQNNAYAV AVANYAPNLS KDPVLSTISK SATTPEPNKK P.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| GABRA2 | |
| Polyclonal | |
| Unconjugated | |
| GABRA2 | |
| C630048P16Rik; FLJ97076; GABA(A) receptor; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; GABAARalpha2; Gabra2; Gabra-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; gamma-aminobutyric acid receptor subunit alpha-2; gamma-aminobutyric acid type A receptor alpha2 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14395, 2555 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.01 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26048, P47869 | |
| GABRA2 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABRA2 (NP_000798.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction