Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533569
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533569 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533569 Supplier Invitrogen™ Supplier No. PA533569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Elephant, Dog, Bat, Bovine, Horse, Rabbit (100%) Opossum, Platypus (87%).

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Gabra3
Gene Accession No. P10064, P20236, P26049, P34903
Gene Alias GABA(A) receptor; GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABA-A receptor alpha-3 subunit; Gabra3; Gabra-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 3; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 3; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 3; gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid receptor, subunit alpha 3; gamma-aminobutyric acid type A receptor alpha3 subunit; gamma-aminobutyric acid type A receptor subunit alpha3; Gamma-animobutyric acid (GABA) A receptor alpha 3; gamma-animobutyric acid (GABA) A receptor, alpha 3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid type A receptor subunit alpha3; MGC33793; unnamed protein product
Gene Symbols Gabra3
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-Terminus of human GABRA3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100063420, 100349107, 14396, 24947, 2556, 282237, 492223
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.