Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546829 Supplier Invitrogen™ Supplier No. PA546829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GTTGQGESRR QEPGDFVKQD IGGLSPKHAP DIPDDSTDNI TIFTRILDRL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gabra3
Gene Accession No. P26049
Gene Alias GABA(A) receptor; GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABA-A receptor alpha-3 subunit; Gabra3; Gabra-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 3; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 3; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 3; gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid receptor, subunit alpha 3; gamma-aminobutyric acid type A receptor alpha3 subunit; gamma-aminobutyric acid type A receptor subunit alpha3; Gamma-animobutyric acid (GABA) A receptor alpha 3; gamma-animobutyric acid (GABA) A receptor, alpha 3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid type A receptor subunit alpha3; MGC33793; unnamed protein product
Gene Symbols Gabra3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GABRA3 (aa 25-74).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14396
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.