Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109752 Supplier Invitrogen™ Supplier No. PA5109752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNQKEEKLCT ENFTRILDSL LDGYDNRLRP GFGGPVTEVK TDIYVTSFGP VSDVEMEYTM DVFFRQTWID KRLKYDGPIE ILRLNNMMVT KVWTPDTFFR NGKKSVSHNM TAPNKLFRIM RNGTILYTMR LTISAECPMR LVDFPMDGHA CPLKFGSYAY PKSEMIYTWT KGPEKSVEVP KESSSLVQYD LIGQTVSSET IKSITGEYIV MTVYFHLRRK MGY.

Gamma-aminobutyric acid is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. This gene encodes subunit alpha-4, which is involved in the etiology of autism and eventually increases autism risk through interaction with another subunit, gamma-aminobutyric acid receptor beta-1. Alternatively spliced transcript variants encoding different isoforms have been found in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GABRA4
Gene Accession No. P28471
Gene Alias GABA(A) receptor; GABA(A) receptor subunit alpha-4; GABA(A) receptor, alpha 4; GABA-A receptor alpha-4 subunit; GABAARalpha4; Gabra4; Gabra-4; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 4; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 4; gamma-aminobutyric acid A receptor alpha 4; gamma-aminobutyric acid receptor subunit alpha-4; gamma-aminobutyric acid type A receptor alpha4 subunit
Gene Symbols GABRA4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 36-258 of human GABRA4 (NP_000800.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140675
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.