Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABRA5 Monoclonal Antibody (OTI1G5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA806598

Description
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described.Specifications
GABRA5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P31644 | |
GABRA5 | |
Human recombit protein fragment corresponding to amino acids 32-259 of human GABRA5 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1G5 | |
Unconjugated | |
GABRA5 | |
gamma-aminobutyric acid type A receptor alpha5 subunit | |
Mouse | |
Affinity Chromatography | |
RUO | |
2558 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction