Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119957 Supplier Invitrogen™ Supplier No. PA5119957
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Jurkat cell lysate, PC-3 cell lysate, rat testis tissue lysates, rat testis tissue, rat brain tissue, mouse brain tissue. Subcellular Location: Postsynaptic cell membrane, Cell membrane.

Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABA-A Receptor (GABA-A R) Subtype. GABA-A Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABA-A R is a multimeric subunit complex. To date six alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha and beta subunits results in the expression of functional GABA-A Rs sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha subunits of the receptor. Lastly, phosphorylation of beta subunits of the receptor has been shown to modulate GABA-A R function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Gabra6
Gene Accession No. P16305, P30191, Q16445
Gene Alias alpha6; GABA subunit A receptor alpha 6; GABA(A) receptor subunit alpha-6; GABA(A) receptor, alpha 6; GABA-A receptor alpha-6 subunit; GABAAR alpha6; GABA-ARalpha6; GABRA 6; Gabra6; Gabra-6; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 6; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 6; gamma-aminobutyric acid A receptor, alpha 6; gamma-aminobutyric acid receptor subunit alpha-6; gamma-aminobutyric acid type A receptor alpha6 subunit; subunit alpha 6 gamma-aminobutyric acid (GABA-A) receptor
Gene Symbols Gabra6
Host Species Rabbit
Immunogen Recombinant protein within human GABRA6 aa 43-242.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14399, 2559, 29708
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.