Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111333 Supplier Invitrogen™ Supplier No. PA5111333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFFGKGPQKK GASKQDQSAN EKNKLEMNKV QVDAHGNI Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 97%.

GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gabrb1
Gene Accession No. P18505
Gene Alias AW061132; B230208N19Rik; GABA beta subunit; GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; Gabrb1; Gabrb-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, subunit beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; gamma-aminobutyric acid type A receptor beta1 subunit; gamma-aminobutyric acid type A receptor subunit beta1; GARB1; GBRB1; si:dkey-20i10.4
Gene Symbols Gabrb1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human GABRB1. Recombinant protein control fragment (Product #RP-101407).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2560
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.