Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRB1 Recombinant Rabbit Monoclonal Antibody (1Q3A9)
GREENER_CHOICE

Catalog No. PIMA537949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537949 Supplier Invitrogen™ Supplier No. MA537949
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: SH-SY5Y, Mouse spinal cord, Mouse brain, Rat brain Immunogen sequence: EKNKLEMNKV QVDAHGNILL STLEIRNETS GSEVLTSVSD PKATMYSYDS ASIQYRKPLS SREAYGRALD RHGVPSKGRI RRRASQLKVK IPDLTDVNSI D.

GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRB1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1Q3A9
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gabrb1
Gene Accession No. P15431, P18505, P50571
Gene Alias AW061132; B230208N19Rik; GABA beta subunit; GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; Gabrb1; Gabrb-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, subunit beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; gamma-aminobutyric acid type A receptor beta1 subunit; gamma-aminobutyric acid type A receptor subunit beta1; GARB1; GBRB1; si:dkey-20i10.4
Gene Symbols Gabrb1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human GABA A Receptor beta 1 (GABRB1) (NP_0008032).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14400, 25450, 2560
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.