Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABRB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564331
Description
Immunogen sequence: STLEIKNEMA TSEAVMGLGD PRSTMLAYDA SSIQYRKAGL PRHSFGRNAL ERHVAQKKSR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA Rα1-6, GABAA R β1-3, GABAA Rγ1-3, GABAA Rδ, GABAA Rε, GABAA Rρ1 and GABAA Rρ2. The GABAB family is composed of GABAB R1α and GABAB R1β. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.
Specifications
GABRB2 | |
Polyclonal | |
Unconjugated | |
GABRB2 | |
AI834970; C030002O17Rik; C030021G16Rik; GABA(A) receptor subunit beta-2; GABAARbeta2; Gabrab2; GABRB2; Gabrb-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 2; gamma-aminobutyric acid (GABA) A receptor, subunit beta 2; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 2; gamma-aminobutyric acid A receptor beta 2; gamma-aminobutyric acid A receptor, subunit beta 2; Gamma-aminobutyric acid receptor beta 2; gamma-aminobutyric acid receptor beta-2 subunit; gamma-aminobutyric acid receptor subunit beta-2; gamma-aminobutyric acid receptor, subunit beta 2; gamma-aminobutyric acid type A receptor beta2 subunit; GARB2; MGC119386; MGC119388; MGC119389 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2561 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P47870 | |
GABRB2 | |
Recombinant protein corresponding to Human GABRB2. Recombinant protein control fragment (Product #RP-104086). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction